3-Amino-2-bromo-5-methylpyridine 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR40336-5GEA
546
DKK
APOSOR40336-5G
3-Amino-2-bromo-5-methylpyridine 98%
3-Amino-2-bromo-5-methylpyridine
Formel:
C₆H₇BrN₂ Molekylvægt: 187,04 g/mol Smeltepunkt: 103…106 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08277281 CAS-nummer: 34552-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...