4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR4023-1GEA
0
DKK
APOSOR4023-1G
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid
Formel:
C₁₀H₁₁BN₂O₃S Molekylvægt: 250,0800 g/mol Smeltepunkt: 248…252 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06659881 CAS-nummer: 850568-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...