3-Chloro-5-nitrobenzaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR401024-1GEA 976 DKK
APOSOR401024-1G APOSOR401024-5G APOSOR401024-10G
3-Chloro-5-nitrobenzaldehyde
3-Chloro-5-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08236780
CAS-nummer: 22233-54-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR