5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR3810-1GEA 143 DKK
APOSOR3810-1G
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid 98%
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid
Formel: C₆H₅ClN₂O₂S
Molekylvægt: 204,6300 g/mol
Smeltepunkt: 204…205 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00173907
CAS-nummer: 61727-33-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR