1-Phenyl-1H-pyrazole-4-carbaldehyde 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR3604-5GEA
0
DKK
APOSOR3604-5G
APOSOR3604-1G
APOSOR3604-10G
1-Phenyl-1H-pyrazole-4-carbaldehyde 98%
1-Phenyl-1H-pyrazole-4-carbaldehyde
Formel:
C₁₀H₈N₂O Molekylvægt: 172,18 g/mol Smeltepunkt: 84…88 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02179568 CAS-nummer: 54605-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...