4-Bromo-1-naphthaleneboronic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR360217-5GEA 688 DKK
APOSOR360217-5G
4-Bromo-1-naphthaleneboronic acid 97%
4-Bromo-1-naphthaleneboronic acid
Formel: C₁₀H₈BBrO₂
Molekylvægt: 250,88
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08701764
CAS-nummer: 145965-14-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR