2-Bromo-N-methylbenzamide 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR3594-1GEA
0
DKK
APOSOR3594-1G
APOSOR3594-5G
2-Bromo-N-methylbenzamide 98%
2-Bromo-N-methylbenzamide
Formel:
C₈H₈BrNO Molekylvægt: 214,06 g/mol Smeltepunkt: 143…146 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00099250 CAS-nummer: 61436-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...