Methyl-3-methoxysalicylate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR350241-50GEA 0 DKK
APOSOR350241-50G
Methyl-3-methoxysalicylate
Methyl-3-methoxysalicylate
Formel: C₉H₁₀O₄
Molekylvægt: 182,1700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00239511
CAS-nummer: 6342-70-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR