(S)-(+)-2-Heptyl isothiocyanate
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR322583-1GEA
0
DKK
APOSOR322583-1G
(S)-(+)-2-Heptyl isothiocyanate
(S)-(+)-2-Heptyl isothiocyanate
Formel:
C₈H₁₅NS Molekylvægt: 157,28 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD05664052 CAS-nummer: 737000-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...