4-(2-Thienyl)aniline

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR322359-5GEA 0 DKK
APOSOR322359-5G APOSOR322359-1G
4-(2-Thienyl)aniline
4-(2-Thienyl)aniline
Formel: C₁₀H₉NS
Molekylvægt: 175,25 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01224064
CAS-nummer: 70010-48-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR