2-(2-Hydroxyethoxy)benzaldehyde
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR322350-10GEA
0
DKK
APOSOR322350-10G
2-(2-Hydroxyethoxy)benzaldehyde
2-(2-Hydroxyethoxy)benzaldehyde
Formel:
C₉H₁₀O₃ Molekylvægt: 166,1739 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00191449 CAS-nummer: 22042-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...