2-Amino-6-bromo-3-formylchromone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR322331-1GEA 1160 DKK
APOSOR322331-1G
2-Amino-6-bromo-3-formylchromone
2-Amino-6-bromo-3-formylchromone
Formel: C₁₀H₆BrNO₃
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 73262-04-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR