Imidazo[1,2-a]pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR322283-1GEA 1120 DKK
APOSOR322283-1G
Imidazo[1,2-a]pyridine
Imidazo[1,2-a]pyridine
Formel: C₇H₆N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005553
CAS-nummer: 274-76-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR