3-(3-Chlorobenzyloxy)phenylboronic acid

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR322261-250MGEA 0 DKK
APOSOR322261-250MG
3-(3-Chlorobenzyloxy)phenylboronic acid
3-(3-Chlorobenzyloxy)phenylboronic acid
Formel: C₁₃H₁₂BClO₃
Molekylvægt: 262,5 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06411352
CAS-nummer: 849062-33-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR