4-Methylphthalic acid

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR322110-25GEA 0 DKK
APOSOR322110-25G
4-Methylphthalic acid
4-Methylphthalic acid
Formel: C₉H₈O₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041946
CAS-nummer: 4316-23-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR