N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR315738-25GEA 0 DKK
APOSOR315738-25G
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide 97%
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
Formel: C₁₄H₂₁N₃O
Molekylvægt: 247,34 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06384973
CAS-nummer: 5294-61-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR