α,α,α,2,4-Pentachlorotoluene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR315021-500GEA 1370 DKK
APOSOR315021-500G
α,α,α,2,4-Pentachlorotoluene
α,α,α,2,4-Pentachlorotoluene
Formel: C₇H₃Cl₅
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00018818
CAS-nummer: 13014-18-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR