1-(4-Bromophenylsulphonyl)-4-methylpiperazine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310452-5GEA 0 DKK
APOSOR310452-5G
1-(4-Bromophenylsulphonyl)-4-methylpiperazine
1-(4-Bromophenylsulphonyl)-4-methylpiperazine
Formel: C₁₁H₁₅BrN₂O₂S
Molekylvægt: 319,2180 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00595522
CAS-nummer: 837-12-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR