5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310199-1GEA 4590 DKK
APOSOR310199-1G APOSOR310199-500MG
5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
5-Bromo-3-methyl-2-(N,2,2,2-tetramethylacetamido)pyridine
Formel: C₁₂H₁₇BrN₂O
Molekylvægt: 285,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09037448
CAS-nummer: 245765-92-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR