6-Amino-5-bromoquinoxaline 95+%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR30436-25GEA
0
DKK
APOSOR30436-25G
6-Amino-5-bromoquinoxaline 95+%
6-Amino-5-bromoquinoxaline
Formel:
C₈H₆BrN₃ Molekylvægt: 224,0573 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00799024 CAS-nummer: 50358-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...