3-Methoxypyridazine 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

APOSOR304308-5GEA
841
DKK
APOSOR304308-5G
3-Methoxypyridazine 97%
3-Methoxypyridazine
Formel:
C₅H₆N₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00234098 CAS-nummer: 19064-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...