3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR303458-1GEA 819 DKK
APOSOR303458-1G APOSOR303458-5G APOSOR303458-500MG
3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester
3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester
Formel: C₁₆H₂₀BNO₃
Molekylvægt: 285,1459 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20275029
CAS-nummer: 1357387-28-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR