4-(Phenylcarbamoyl)phenylboronic acid pinacol ester

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303436-5GEA 0 DKK
APOSOR303436-5G APOSOR303436-1G
4-(Phenylcarbamoyl)phenylboronic acid pinacol ester
4-(Phenylcarbamoyl)phenylboronic acid pinacol ester
Formel: C₁₉H₂₂BNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12546047
CAS-nummer: 949115-03-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR