3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR303416-5GEA 2450 DKK
APOSOR303416-5G APOSOR303416-500MG APOSOR303416-1G
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
3-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Formel: C₁₈H₂₇BN₂O₃
Molekylvægt: 330,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05864308
CAS-nummer: 883738-38-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR