3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR303408-10GEA
0
DKK
APOSOR303408-10G
APOSOR303408-5G
APOSOR303408-1G
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
3-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Formel:
C₁₇H₂₄BNO₄ Molekylvægt: 317,19 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09266172 CAS-nummer: 1036991-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...