6-Bromopyrazolo[1,5-a]pyrimidine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSOR303202-1GEA 656 DKK
APOSOR303202-1G APOSOR303202-5G APOSOR303202-25G
6-Bromopyrazolo[1,5-a]pyrimidine
6-Bromopyrazolo[1,5-a]pyrimidine
Formel: C₆H₄BrN₃
Molekylvægt: 198,02 g/mol
Smeltepunkt: 122…124 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09832894
CAS-nummer: 705263-10-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR