7-Chloroimidazo[4,5-b]pyridine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR301113-1GEA
775
DKK
APOSOR301113-1G
7-Chloroimidazo[4,5-b]pyridine
7-Chloroimidazo[4,5-b]pyridine
Formel:
C₆H₄ClN₃ Molekylvægt: 153,5691 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD15143399 CAS-nummer: 6980-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...