Ethyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR300507-1GEA 524 DKK
APOSOR300507-1G
Ethyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₀H₉BrN₂O₂
Molekylvægt: 269,1000 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10698068
CAS-nummer: 372198-69-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR