4-tert-Butyl-3-thiosemicarbazide
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR29873-5GEA
0
DKK
APOSOR29873-5G
APOSOR29873-1G
4-tert-Butyl-3-thiosemicarbazide
4-tert-Butyl-3-thiosemicarbazide
Formel:
C₅H₁₃N₃S Molekylvægt: 147,24 g/mol Smeltepunkt: 139…140 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00041304 CAS-nummer: 13431-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...