6-Methylnicotinamide 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering

|
Advarsel
|
APOSOR29753-10GEA
524
DKK
APOSOR29753-10G
APOSOR29753-25G
6-Methylnicotinamide 98%
6-Methylnicotinamide
Formel:
C₇H₈N₂O Molekylvægt: 136,15 Smeltepunkt: 196…198 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006342 CAS-nummer: 6960-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...