3,5-Dimethyl-1H-pyrazole-1-methanol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR28784-5GEA
197
DKK
APOSOR28784-5G
3,5-Dimethyl-1H-pyrazole-1-methanol
3,5-Dimethyl-1H-pyrazole-1-methanol
Formel:
C₆H₁₀N₂O Smeltepunkt: 107…112 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00020738 CAS-nummer: 85264-33-1 EINECS: 286-553-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...