2-(2-Thienyl)-4-thiazolecarboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR23153-1GEA 1760 DKK
APOSOR23153-1G
2-(2-Thienyl)-4-thiazolecarboxylic acid
2-(2-Thienyl)-4-thiazolecarboxylic acid
Formel: C₈H₅NO₂S₂
Molekylvægt: 211,25
Smeltepunkt: 156…160 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01936014
CAS-nummer: 24044-07-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR