6-Amino-2-methylquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR21398-1GEA 765 DKK
APOSOR21398-1G
6-Amino-2-methylquinoline
6-Amino-2-methylquinoline
Formel: C₁₀H₁₀N₂
Molekylvægt: 158,20 g/mol
Smeltepunkt: 189…191 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052600
CAS-nummer: 65079-19-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR