1-Bromo-4-(trans-4-propylcyclohexyl)benzene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR16675-5GEA 0 DKK
APOSOR16675-5G APOSOR16675-25G APOSOR16675-100G
1-Bromo-4-(trans-4-propylcyclohexyl)benzene 98%
1-Bromo-4-(trans-4-propylcyclohexyl)benzene
Formel: C₁₅H₂₁Br
Molekylvægt: 281,2300 g/mol
Smeltepunkt: 51…52 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00577367
CAS-nummer: 86579-53-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR