2-Boc-aminothiazole-5-carboxylic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR15913-5GEA 1110 DKK
APOSOR15913-5G
2-Boc-aminothiazole-5-carboxylic acid 97%
2-Boc-aminothiazole-5-carboxylic acid
Formel: C₉H₁₂N₂O₄S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06796614
CAS-nummer: 302964-02-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR