4-Amino-5-bromo-2-chloropyrimidine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR15511-1GEA
110
DKK
APOSOR15511-1G
4-Amino-5-bromo-2-chloropyrimidine
4-Amino-5-bromo-2-chloropyrimidine
Formel:
C₄H₃BrClN₃ Molekylvægt: 208,44 g/mol Smeltepunkt: 189…191 °C Opbevaringstemperatur: Stue temperatur |
CAS-nummer:
205672-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...