5-Amino-3-(4-chlorophenyl)isoxazole
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR15282-1GEA
0
DKK
APOSOR15282-1G
APOSOR15282-5G
5-Amino-3-(4-chlorophenyl)isoxazole
5-Amino-3-(4-chlorophenyl)isoxazole
Formel:
C₉H₇ClN₂O Molekylvægt: 194,6177 g/mol Smeltepunkt: 162…165 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01569450 CAS-nummer: 33866-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...