4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR14938-1GEA 2670 DKK
APOSOR14938-1G
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde 97%
4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde
Formel: C₁₂H₁₂N₂O
Smeltepunkt: 78…81 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06800625
CAS-nummer: 934570-54-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR