Ethyl imidazo[1,2-a]pyridine-2-carboxylate 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR1446-1GEA
0
DKK
APOSOR1446-1G
APOSOR1446-5G
Ethyl imidazo[1,2-a]pyridine-2-carboxylate 97%
Ethyl imidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₁₀N₂O₂ Molekylvægt: 190,20 g/mol Smeltepunkt: 79…87 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02325394 CAS-nummer: 38922-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...