2-Phenylisonicotinonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


APOSOR14232-5GEA 7920 DKK
APOSOR14232-5G APOSOR14232-1G
2-Phenylisonicotinonitrile
2-Phenylisonicotinonitrile
Formel: C₁₂H₈N₂
Smeltepunkt: 78…80 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06200958
CAS-nummer: 33744-17-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR