3-Pyrid-3-ylbenzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR1409-1GEA 0 DKK
APOSOR1409-1G APOSOR1409-5G
3-Pyrid-3-ylbenzoic acid 98%
3-Pyrid-3-ylbenzoic acid
Formel: C₁₂H₉NO₂
Molekylvægt: 199,2054 g/mol
Smeltepunkt: 202…206 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03426512
CAS-nummer: 4385-77-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR