Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR11996-1GEA 898 DKK
APOSOR11996-1G
Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₁H₁₂N₂O₂
Molekylvægt: 204,2300 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02325396
CAS-nummer: 70705-30-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR