4-Bromo-N-cyclopentylbenzamide
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR11627-5GEA
0
DKK
APOSOR11627-5G
APOSOR11627-1G
4-Bromo-N-cyclopentylbenzamide
4-Bromo-N-cyclopentylbenzamide
Formel:
C₁₂H₁₄BrNO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00588998 CAS-nummer: 223557-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...