2-(Methoxycarbonyl)phenylboronic acid pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR11608-1GEA 427 DKK
APOSOR11608-1G
2-(Methoxycarbonyl)phenylboronic acid pinacol ester
2-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₄
Molekylvægt: 262,1093 g/mol
Smeltepunkt: 42…44 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05663866
CAS-nummer: 653589-95-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR