2,3,5,6-Tetramethyl-m-phenylendiamin

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR11241-25GEA 0 DKK
APOSOR11241-25G APOSOR11241-5G
2,3,5,6-Tetramethyl-m-phenylendiamin
2,3,5,6-Tetramethyl-m-phenylendiamin
Formel: C₁₀H₁₆N₂
Molekylvægt: 164,25 g/mol
Smeltepunkt: 150…155 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007905
CAS-nummer: 3102-87-2
EINECS: 221-457-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR