5-Chlorobenzimidazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
APOSOR11144-1GEA
132
DKK
APOSOR11144-1G
5-Chlorobenzimidazole
5-Chlorobenzimidazole
Formel:
C₇H₅ClN₂ Molekylvægt: 152,5810 g/mol Smeltepunkt: 118…122 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD17015012 CAS-nummer: 4887-82-5 EINECS: 225-503-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...