Imidazo[1,2-a]pyridine-8-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR110823-1GEA 186 DKK
APOSOR110823-1G
Imidazo[1,2-a]pyridine-8-carboxylic acid
Imidazo[1,2-a]pyridine-8-carboxylic acid
Formel: C₈H₆N₂O₂
Molekylvægt: 162,1454 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06739251
CAS-nummer: 133427-08-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR