1-Boc-4-(4-nitrophenyl)piperazine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR110377-5GEA 524 DKK
APOSOR110377-5G
1-Boc-4-(4-nitrophenyl)piperazine 95%
1-Boc-4-(4-nitrophenyl)piperazine
Formel: C₁₅H₂₁N₃O₄
Molekylvægt: 307,34 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02187326
CAS-nummer: 182618-86-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR