2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR10381-1GEA 668 DKK
APOSOR10381-1G APOSOR10381-250MG
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₃H₁₈BBrO₂
Molekylvægt: 297,00
Smeltepunkt: 85…89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02179493
CAS-nummer: 138500-85-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR