3-(Indol-3-yl)propionsyre 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR10180-100GEA 0 DKK
APOSOR10180-100G
3-(Indol-3-yl)propionsyre 97%
3-(Indol-3-yl)propionsyre
Formel: C₁₁H₁₁NO₂
Molekylvægt: 189,21 g/mol
Smeltepunkt: 134…135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005660
CAS-nummer: 830-96-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR